Product Details
- SNP ID
-
rs112114803
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:54283626 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAACTATACAGAGCGATTTTTTT[A/T]ATACAATTATTACAACGATTAAAAA
- Phenotype
-
MIM: 612985
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IRX3
PubMed Links
Gene Details
- Gene
- IRX3
- Gene Name
- iroquois homeobox 3
View Full Product Details