Product Details
- SNP ID
-
rs116380761
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:17482651 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTCAGATTTGTTAGGCCTTTCCC[G/T]TATGGCTTCTGAGTTTTGGTCCTAC
- Phenotype
-
MIM: 609878
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MED9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199913278] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MED9
- Gene Name
- mediator complex subunit 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018019.2 |
|
Intron |
|
|
NP_060489.1 |
View Full Product Details