Product Details

SNP ID
rs112038108
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4900468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCTGCCTCATTCCTGCGACAG[C/T]CGCAACAGCAGCTAGGCCTCGGAAT
Phenotype
MIM: 100725 MIM: 609426
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C17orf107 PubMed Links
Additional Information
For this assay, SNP(s) [rs35400274] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C17orf107
Gene Name
chromosome 17 open reading frame 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145536.1 618 Missense Mutation CCG,TCG P170S NP_001139008.1
Gene
CHRNE
Gene Name
cholinergic receptor nicotinic epsilon subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000080.3 618 Intron NP_000071.1
XM_017024115.1 618 Intron XP_016879604.1
Gene
MINK1
Gene Name
misshapen like kinase 1
There are no transcripts associated with this gene.

View Full Product Details