Product Details

SNP ID
rs116480053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38805358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGCAACATATTTGTATTTTTA[A/G]GAATATGTAGCTATGAAAAGAAACT
Phenotype
MIM: 603261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CWC25 PubMed Links

Gene Details

Gene
CWC25
Gene Name
CWC25 spliceosome associated protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017748.4 Intron NP_060218.1
Gene
PIP4K2B
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 beta
There are no transcripts associated with this gene.

View Full Product Details