Product Details

SNP ID
rs117771322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40189318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGACGGAGAAACTTCAATATTCA[C/G]AGGAGCCCGCGGGGCGTGAGGATTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6867 PubMed Links

Gene Details

Gene
MIR6867
Gene Name
microRNA 6867
There are no transcripts associated with this gene.

Gene
RAPGEFL1
Gene Name
Rap guanine nucleotide exchange factor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303533.1 317 Missense Mutation CAG,GAG Q202E NP_001290462.1
NM_001303534.1 317 Missense Mutation CAG,GAG Q196E NP_001290463.1
NM_016339.4 317 Missense Mutation CAG,GAG Q147E NP_057423.1
XM_006721932.3 317 Missense Mutation CAG,GAG Q353E XP_006721995.1
XM_006721935.3 317 Missense Mutation CAG,GAG Q184E XP_006721998.1
XM_006721937.3 317 Missense Mutation CAG,GAG Q147E XP_006722000.1
XM_006721938.3 317 Missense Mutation CAG,GAG Q83E XP_006722001.1

View Full Product Details