Product Details

SNP ID
rs117953693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74951031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATGCCCCAAGGATACGGGAGG[A/T]GCCCCAGTCTGGCTTTTGCAGTCGG
Phenotype
MIM: 605752 MIM: 607828
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HID1 PubMed Links

Gene Details

Gene
HID1
Gene Name
HID1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030630.2 3050 UTR 3 NP_085133.1
XM_005257226.1 3050 UTR 3 XP_005257283.1
Gene
OTOP3
Gene Name
otopetrin 3
There are no transcripts associated with this gene.

View Full Product Details