Product Details

SNP ID
rs115919790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75946274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAATAAGTTTCCTTCCCCCAGTC[C/T]CTTTTCTTCAATCCTGCTTTTAAGC
Phenotype
MIM: 609751 MIM: 616807
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOX1 PubMed Links

Gene Details

Gene
ACOX1
Gene Name
acyl-CoA oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185039.1 2818 UTR 3 NP_001171968.1
NM_004035.6 2818 UTR 3 NP_004026.2
NM_007292.5 2818 UTR 3 NP_009223.2
XM_011524868.2 2818 UTR 3 XP_011523170.1
XM_011524869.2 2818 UTR 3 XP_011523171.1
Gene
FBF1
Gene Name
Fas binding factor 1
There are no transcripts associated with this gene.

View Full Product Details