Product Details

SNP ID
rs113889392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28043303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGATATTGAGCCGGATAGACCT[A/G]TTGGATATGGAGCCTTTGGTGTTGT
Phenotype
MIM: 609476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLK PubMed Links

Gene Details

Gene
NLK
Gene Name
nemo like kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016231.4 657 Missense Mutation ATT,GTT I144V NP_057315.3
XM_005257988.2 657 Missense Mutation ATT,GTT I144V XP_005258045.1

View Full Product Details