Product Details

SNP ID
rs116860247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12449379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAGGAAGTAGCTACTGGCTTCC[A/G]AAGCCACACACACACAAAAGTAAGT
Phenotype
MIM: 609216
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371998 PubMed Links

Gene Details

Gene
LOC105371998
Gene Name
uncharacterized LOC105371998
There are no transcripts associated with this gene.

Gene
SPIRE1
Gene Name
spire type actin nucleation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128626.1 2940 UTR 3 NP_001122098.1
NM_001128627.1 2940 UTR 3 NP_001122099.1
NM_020148.2 2940 UTR 3 NP_064533.3
XM_005258122.4 2940 UTR 3 XP_005258179.1
XM_011525701.1 2940 UTR 3 XP_011524003.1
XM_011525702.1 2940 UTR 3 XP_011524004.1
XM_011525703.2 2940 Intron XP_011524005.1

View Full Product Details