Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001289036.1 | 197 | Intron | NP_001275965.1 | ||
NM_001318522.1 | 197 | Intron | NP_001305451.1 | ||
NM_014410.4 | 197 | Intron | NP_055225.1 | ||
NM_199167.1 | 197 | Intron | NP_954636.1 | ||
XM_005258103.3 | 197 | Intron | XP_005258160.1 | ||
XM_006722322.2 | 197 | Intron | XP_006722385.1 | ||
XM_011525648.2 | 197 | Intron | XP_011523950.1 | ||
XM_011525649.1 | 197 | Intron | XP_011523951.1 | ||
XM_011525651.2 | 197 | UTR 5 | XP_011523953.1 | ||
XM_011525652.1 | 197 | Intron | XP_011523954.1 | ||
XM_011525653.2 | 197 | Intron | XP_011523955.1 | ||
XM_017025707.1 | 197 | Intron | XP_016881196.1 |