Product Details

SNP ID
rs115490373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48341133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGCCAGCAGGTGAATGATCGC[A/G]TACGACAGATGCTTCACACCAGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM143 PubMed Links

Gene Details

Gene
TMEM143
Gene Name
transmembrane protein 143
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303538.1 Intron NP_001290467.1
NM_001303539.1 Intron NP_001290468.1
NM_001303540.1 Intron NP_001290469.1
NM_018273.3 Intron NP_060743.2

View Full Product Details