Product Details

SNP ID
rs116177062
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48489070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACTTCTAAGGGGAAAACAAAGC[C/T]CTTTCCTCTTGTCATCTGCTTTCTC
Phenotype
MIM: 602488
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CYTH2 PubMed Links

Gene Details

Gene
CYTH2
Gene Name
cytohesin 2
There are no transcripts associated with this gene.

Gene
LMTK3
Gene Name
lemur tyrosine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080434.1 Intron NP_001073903.1
XM_011526411.2 Intron XP_011524713.1
XM_011526412.2 Intron XP_011524714.1

View Full Product Details