Product Details
- SNP ID
-
rs115663096
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58127937 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGATGTCGAACAAGGGATGAGC[C/T]CTTTTGGAACATTTTTCCACACTGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF329
PubMed Links
Gene Details
- Gene
- ZNF329
- Gene Name
- zinc finger protein 329
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024620.3 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
NP_078896.3 |
XM_006723381.2 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_006723444.1 |
XM_006723382.3 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_006723445.1 |
XM_006723383.3 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_006723446.1 |
XM_006723384.3 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_006723447.1 |
XM_011527310.2 |
1804 |
Missense Mutation |
AGC,GGC |
S546G |
XP_011525612.1 |
XM_011527311.2 |
1804 |
Missense Mutation |
AGC,GGC |
S546G |
XP_011525613.1 |
XM_011527312.2 |
1804 |
Missense Mutation |
AGC,GGC |
S546G |
XP_011525614.1 |
XM_011527313.2 |
1804 |
Missense Mutation |
AGC,GGC |
S546G |
XP_011525615.1 |
XM_011527314.2 |
1804 |
Missense Mutation |
AGC,GGC |
S546G |
XP_011525616.1 |
XM_011527315.1 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_011525617.1 |
XM_011527316.2 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_011525618.1 |
XM_017027307.1 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_016882796.1 |
XM_017027308.1 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_016882797.1 |
XM_017027309.1 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_016882798.1 |
XM_017027310.1 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_016882799.1 |
XM_017027311.1 |
1804 |
Missense Mutation |
AGC,GGC |
S523G |
XP_016882800.1 |
- Gene
- ZSCAN18
- Gene Name
- zinc finger and SCAN domain containing 18
There are no transcripts associated with this gene.
View Full Product Details