Product Details

SNP ID
rs115396898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58349891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTACGACCTCAAGATCTGGAG[A/G]TCAGACCTGTGCCCTCACGCTCTGC
Phenotype
MIM: 138670 MIM: 165260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
A1BG PubMed Links

Gene Details

Gene
A1BG
Gene Name
alpha-1-B glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130786.3 Intron NP_570602.2
Gene
A1BG-AS1
Gene Name
A1BG antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC100419840
Gene Name
zinc finger protein 446 pseudogene
There are no transcripts associated with this gene.

Gene
ZNF497
Gene Name
zinc finger protein 497
There are no transcripts associated with this gene.

Gene
ZSCAN22
Gene Name
zinc finger and SCAN domain containing 22
There are no transcripts associated with this gene.

View Full Product Details