Product Details

SNP ID
rs116528776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33300211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATCAAAAGTAATCCCAAAACC[A/C]AAAGGAAAGGGAGTCTCAGACCCTT
Phenotype
MIM: 116897
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEBPA PubMed Links

Gene Details

Gene
CEBPA
Gene Name
CCAAT/enhancer binding protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285829.1 2354 UTR 3 NP_001272758.1
NM_001287424.1 2354 UTR 3 NP_001274353.1
NM_001287435.1 2354 UTR 3 NP_001274364.1
NM_004364.4 2354 UTR 3 NP_004355.2
Gene
CEBPA-AS1
Gene Name
CEBPA antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details