Product Details

SNP ID
rs112865072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35778511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTACGATGACTTTCGTTCCCT[C/G]GATGCCCACCTCCACCGGTGCATAT
Phenotype
MIM: 614902
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP33 PubMed Links

Gene Details

Gene
ARHGAP33
Gene Name
Rho GTPase activating protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172630.1 1858 UTR 5 NP_001166101.1
NM_052948.3 1858 Silent Mutation CTC,CTG L106L NP_443180.2
XM_005258488.1 1858 Silent Mutation CTC,CTG L106L XP_005258545.1
XM_006722999.1 1858 Silent Mutation CTC,CTG L106L XP_006723062.1
XM_006723003.2 1858 Intron XP_006723066.1
XM_011526416.1 1858 UTR 5 XP_011524718.1
XM_011526417.1 1858 UTR 5 XP_011524719.1
XM_011526418.1 1858 UTR 5 XP_011524720.1
XM_011526419.1 1858 UTR 5 XP_011524721.1
XM_011526420.1 1858 UTR 5 XP_011524722.1
XM_011526421.2 1858 Silent Mutation CTC,CTG L143L XP_011524723.1
XM_017026238.1 1858 Silent Mutation CTC,CTG L143L XP_016881727.1
Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

View Full Product Details