Product Details

SNP ID
rs114482500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2850611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCAGTGGTCTTTGAGGATGTG[A/G]CTGTGGACTTCACCCTGGAGGAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF555 PubMed Links

Gene Details

Gene
ZNF555
Gene Name
zinc finger protein 555
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172775.1 169 Missense Mutation ACT,GCT T10A NP_001166246.1
NM_152791.4 169 Missense Mutation ACT,GCT T10A NP_690004.4
XM_011527716.2 169 Missense Mutation ACT,GCT T12A XP_011526018.1
XM_017026375.1 169 Missense Mutation ACT,GCT T12A XP_016881864.1

View Full Product Details