Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001245002.1 | Intron | NP_001231931.1 | |||
NM_001245004.1 | Intron | NP_001231933.1 | |||
NM_001245005.1 | Intron | NP_001231934.1 | |||
NM_005597.3 | Intron | NP_005588.2 | |||
NM_205843.2 | Intron | NP_995315.1 | |||
XM_005259564.3 | Intron | XP_005259621.1 | |||
XM_006722759.2 | Intron | XP_006722822.2 | |||
XM_017026834.1 | Intron | XP_016882323.1 | |||
XM_017026835.1 | Intron | XP_016882324.1 | |||
XM_017026836.1 | Intron | XP_016882325.1 |