Product Details

SNP ID
rs116429568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:686965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGGCTGTCCCCACTGCGGGTG[C/T]AGAGCATGGTAAGTGTCAGGTGGCC
Phenotype
MIM: 605343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
There are no transcripts associated with this gene.

Gene
PRSS57
Gene Name
protease, serine 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308209.1 633 Missense Mutation TAC,TGC Y201C NP_001295138.1
NM_214710.4 633 Missense Mutation TAC,TGC Y202C NP_999875.1

View Full Product Details