Product Details

SNP ID
rs112570154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:931882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGTGCCGTGGAGGGGCAGTGGC[A/G]AGGGTGGGGCGGATTTCAGATGGCG
Phenotype
MIM: 603265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID3A PubMed Links

Gene Details

Gene
ARID3A
Gene Name
AT-rich interaction domain 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005224.2 Intron NP_005215.1
XM_005259513.4 Intron XP_005259570.1
XM_005259514.4 Intron XP_005259571.1
XM_017026445.1 Intron XP_016881934.1
XM_017026446.1 Intron XP_016881935.1
XM_017026447.1 Intron XP_016881936.1

View Full Product Details