Product Details

SNP ID
rs111464692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11298171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCATCATCCTAGTTGGCCTGGG[C/T]ATTGGTGGTAAATGTGGAGGGGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPC102 PubMed Links

Gene Details

Gene
HSPC102
Gene Name
uncharacterized HSPC102
There are no transcripts associated with this gene.

Gene
TSPAN16
Gene Name
tetraspanin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282509.1 189 Silent Mutation GGC,GGT G33G NP_001269438.1
NM_001282510.1 189 Silent Mutation GGC,GGT G33G NP_001269439.1
NM_012466.3 189 Silent Mutation GGC,GGT G33G NP_036598.1
XM_011527901.1 189 Silent Mutation GGC,GGT G33G XP_011526203.1
XM_011527902.1 189 Silent Mutation GGC,GGT G33G XP_011526204.1
XM_011527903.2 189 UTR 5 XP_011526205.1

View Full Product Details