Product Details

SNP ID
rs113581497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12691082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGAGTATTTGGTTAAGTGACTA[C/T]GACCCCAGCCAGGCTGTGAAGCCCC
Phenotype
MIM: 600944 MIM: 609074 MIM: 603002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHPS PubMed Links

Gene Details

Gene
DHPS
Gene Name
deoxyhypusine synthase
There are no transcripts associated with this gene.

Gene
FBXW9
Gene Name
F-box and WD repeat domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032301.2 Intron NP_115677.2
XM_005260096.4 Intron XP_005260153.1
Gene
LOC105372280
Gene Name
uncharacterized LOC105372280
There are no transcripts associated with this gene.

Gene
TNPO2
Gene Name
transportin 2
There are no transcripts associated with this gene.

View Full Product Details