Product Details

SNP ID
rs112963788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14032402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCCAGGCAGCGCCGGGCCACT[A/G]CAGTGCTACGGAGTTGGACCCTTGG
Phenotype
MIM: 605350 MIM: 606855
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004843.3 250 Silent Mutation CTA,CTG L39L NP_004834.1
Gene
RLN3
Gene Name
relaxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311197.1 250 Intron NP_001298126.1
NM_080864.3 250 Intron NP_543140.1

View Full Product Details