Product Details

SNP ID
rs113617557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15111630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCAGAGGGGACCCTGTGTTCAC[A/T]GGCATCCTCAAGGATTATCTTCGAG
Phenotype
MIM: 605770
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ILVBL PubMed Links

Gene Details

Gene
ILVBL
Gene Name
ilvB acetolactate synthase like
There are no transcripts associated with this gene.

Gene
SYDE1
Gene Name
synapse defective Rho GTPase homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300910.1 Intron NP_001287839.1
NM_033025.5 Intron NP_149014.3

View Full Product Details