Product Details

SNP ID
rs114011915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9324205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCATAACGGCCGCCATCTTAAC[A/G]GCGCGTTCCCGTTGGCGTCTGAGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF559 PubMed Links

Gene Details

Gene
ZNF559
Gene Name
zinc finger protein 559
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202406.1 434 Silent Mutation ACA,ACG T9T NP_001189335.1
NM_001202407.1 434 UTR 5 NP_001189336.1
NM_001202408.1 434 Silent Mutation ACA,ACG T9T NP_001189337.1
NM_001202409.1 434 UTR 5 NP_001189338.1
NM_001202410.1 434 UTR 5 NP_001189339.1
NM_001202411.1 434 UTR 5 NP_001189340.1
NM_001202412.1 434 Intron NP_001189341.1
NM_032497.2 434 UTR 5 NP_115886.1
Gene
ZNF559-ZNF177
Gene Name
ZNF559-ZNF177 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172650.2 434 Intron NP_001166121.1
NM_001202425.1 434 Intron NP_001189354.1

View Full Product Details