Product Details

SNP ID
rs116262991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18208133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGGAAGGAAATGCACCAAGTTC[A/G]TGAACCCGGATGACTTTTAGTTCTG
Phenotype
MIM: 600128 MIM: 179490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102725254 PubMed Links

Gene Details

Gene
LOC102725254
Gene Name
uncharacterized LOC102725254
There are no transcripts associated with this gene.

Gene
PDE4C
Gene Name
phosphodiesterase 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011528056.2 5183 Intron XP_011526358.1
XM_011528058.2 5183 Intron XP_011526360.1
XM_017026866.1 5183 UTR 3 XP_016882355.1
Gene
RAB3A
Gene Name
RAB3A, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details