Product Details

SNP ID
rs117421936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62844962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCAAGTCCAGGAGGACACAAG[C/T]GCCATGCTGCCATGGATTTAACTTC
Phenotype
MIM: 120270 MIM: 604745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL9A3 PubMed Links

Gene Details

Gene
COL9A3
Gene Name
collagen type IX alpha 3
There are no transcripts associated with this gene.

Gene
DPH3P1
Gene Name
diphthamide biosynthesis 3 pseudogene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080750.4 Intron NP_542788.1
Gene
TCFL5
Gene Name
transcription factor like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301726.1 Intron NP_001288655.1
NM_006602.3 Intron NP_006593.2
XM_005260185.3 Intron XP_005260242.1
XM_011528497.2 Intron XP_011526799.1
XM_011528498.2 Intron XP_011526800.1
XM_011528499.1 Intron XP_011526801.1

View Full Product Details