Product Details
- SNP ID
-
rs112977482
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63244053 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCTCTTCCTCCGTAAACACGCTG[A/G]CCACCTGGCAGCCACAGACAAAGCC
- Phenotype
-
MIM: 605737
MIM: 612873
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BIRC7
PubMed Links
Gene Details
- Gene
- BIRC7
- Gene Name
- baculoviral IAP repeat containing 7
There are no transcripts associated with this gene.
- Gene
- FLJ16779
- Gene Name
- uncharacterized LOC100192386
There are no transcripts associated with this gene.
- Gene
- LOC107985425
- Gene Name
- uncharacterized LOC107985425
- Gene
- MIR3196
- Gene Name
- microRNA 3196
There are no transcripts associated with this gene.
- Gene
- NKAIN4
- Gene Name
- Na+/K+ transporting ATPase interacting 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152864.3 |
1013 |
Missense Mutation |
GCC,GTC |
A168V |
NP_690603.3 |
XM_005260192.1 |
1013 |
Missense Mutation |
GCC,GTC |
A168V |
XP_005260249.1 |
XM_005260194.3 |
1013 |
Missense Mutation |
GCC,GTC |
A106V |
XP_005260251.1 |
XM_011528527.1 |
1013 |
Missense Mutation |
GCC,GTC |
A168V |
XP_011526829.1 |
XM_011528528.2 |
1013 |
Missense Mutation |
GCC,GTC |
A106V |
XP_011526830.1 |
XM_011528529.2 |
1013 |
Missense Mutation |
GCC,GTC |
A106V |
XP_011526831.1 |
XM_017027636.1 |
1013 |
Missense Mutation |
GCC,GTC |
A106V |
XP_016883125.1 |
XM_017027637.1 |
1013 |
Missense Mutation |
GCC,GTC |
A106V |
XP_016883126.1 |
XM_017027638.1 |
1013 |
Missense Mutation |
GCC,GTC |
A106V |
XP_016883127.1 |
XM_017027639.1 |
1013 |
Missense Mutation |
GCC,GTC |
A106V |
XP_016883128.1 |
View Full Product Details