Product Details

SNP ID
rs113580409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44196972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGAGATGTCTTCTCACACAAA[A/G]TGGCCACAAGGTCTGCCATTAACTA
Phenotype
MIM: 605267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JPH2 PubMed Links

Gene Details

Gene
JPH2
Gene Name
junctophilin 2
There are no transcripts associated with this gene.

Gene
OSER1
Gene Name
oxidative stress responsive serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016470.7 1255 UTR 3 NP_057554.4
XM_011528851.1 1255 UTR 3 XP_011527153.1
XM_011528853.2 1255 UTR 3 XP_011527155.1
XM_011528854.2 1255 UTR 3 XP_011527156.1
XM_017027873.1 1255 UTR 3 XP_016883362.1

View Full Product Details