Product Details

SNP ID
rs115211941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:47507083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGTTGGCGTGGTAGATGGATAG[A/G]TGGGAAAGGACTGCAGAAGTCAGCA
Phenotype
MIM: 601937
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCOA3 PubMed Links

Gene Details

Gene
NCOA3
Gene Name
nuclear receptor coactivator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174087.1 Intron NP_001167558.1
NM_001174088.1 Intron NP_001167559.1
NM_006534.3 Intron NP_006525.2
NM_181659.2 Intron NP_858045.1

View Full Product Details