Product Details

SNP ID
rs118047173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59018389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGTACCCAGCCTGCTGCCACTC[A/C]TGTACACCGCATGAGGGAATGCTTT
Phenotype
MIM: 612901
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TUBB1 PubMed Links

Gene Details

Gene
TUBB1
Gene Name
tubulin beta 1 class VI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030773.3 Intron NP_110400.1
XM_017028085.1 Intron XP_016883574.1

View Full Product Details