Product Details

SNP ID
rs113102870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36614393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGAGCGGGCCAGCCCTCTGCA[A/G]GACACAGGGCTCCTTGAGTAGGCAG
Phenotype
MIM: 606577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf24 PubMed Links

Gene Details

Gene
C20orf24
Gene Name
chromosome 20 open reading frame 24
There are no transcripts associated with this gene.

Gene
SLA2
Gene Name
Src like adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032214.3 1000 Silent Mutation CTG,TTG L193L NP_115590.1
NM_175077.2 1000 Intron NP_778252.1
XM_017028098.1 1000 Intron XP_016883587.1
Gene
TGIF2-C20orf24
Gene Name
TGIF2-C20orf24 readthrough
There are no transcripts associated with this gene.

View Full Product Details