Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152503.5 | Intron | NP_689716.4 | |||
NM_213631.2 | Intron | NP_998796.1 | |||
NM_213632.2 | Intron | NP_998797.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135771.2 | Intron | NP_001129243.1 | |||
NM_001324299.1 | Intron | NP_001311228.1 | |||
NM_001324301.1 | Intron | NP_001311230.1 | |||
NM_001324302.1 | Intron | NP_001311231.1 | |||
NM_001324303.1 | Intron | NP_001311232.1 | |||
NM_001324304.1 | Intron | NP_001311233.1 | |||
NM_001324305.1 | Intron | NP_001311234.1 | |||
NM_001324306.1 | Intron | NP_001311235.1 | |||
NM_002951.4 | Intron | NP_002942.2 | |||
XM_006723851.2 | Intron | XP_006723914.1 | |||
XM_006723852.2 | Intron | XP_006723915.1 |