Product Details

SNP ID
rs111382388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31946480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTCCCCTTCTTATCTCCAGTC[C/T]CTGCTAAGCCAATACCTTGGGCCTC
Phenotype
MIM: 610789
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDRG1 PubMed Links

Gene Details

Gene
PDRG1
Gene Name
p53 and DNA damage regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030815.2 Intron NP_110442.1
Gene
TTLL9
Gene Name
tubulin tyrosine ligase like 9
There are no transcripts associated with this gene.

View Full Product Details