Product Details
- SNP ID
-
rs112910952
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:17944990 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCTGTCTTGCAGTTTTACTTGA[C/G]TAAAACTTTGACCATCACCCAACCT
- Phenotype
-
MIM: 605937
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SNX5
PubMed Links
Gene Details
- Gene
- SNX5
- Gene Name
- sorting nexin 5
View Full Product Details