Product Details

SNP ID
rs111560893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30430285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATCTCTCATCAGCATGTCCTAC[A/T]ACTGCTGCTCTAGAAACTTCTCCTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP13-3 PubMed Links

Gene Details

Gene
KRTAP13-3
Gene Name
keratin associated protein 13-3
There are no transcripts associated with this gene.

Gene
KRTAP13-4
Gene Name
keratin associated protein 13-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181600.1 10 Missense Mutation AAC,TAC N4Y NP_853631.1

View Full Product Details