Product Details

SNP ID
rs118040540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:18906991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCCCCACCCCCTTCCAGGCCT[A/G]ATGGCCCTAGGCCTCCGTGCTTGGT
Phenotype
MIM: 601279 MIM: 606810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGCR6 PubMed Links

Gene Details

Gene
DGCR6
Gene Name
DiGeorge syndrome critical region gene 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005675.4 Intron NP_005666.2
XM_011530394.1 Intron XP_011528696.1
Gene
PRODH
Gene Name
proline dehydrogenase 1
There are no transcripts associated with this gene.

View Full Product Details