Product Details

SNP ID
rs113704375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36564597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGAGTGTGAGGGCTCCGTGGA[A/G]CGCGAGCTGGAGCGGCTGCGGCGCT
Phenotype
MIM: 602911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNG2 PubMed Links

Gene Details

Gene
CACNG2
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006078.3 592 Silent Mutation NP_006069.1
XM_017028531.1 592 Silent Mutation XP_016884020.1

View Full Product Details