Product Details

SNP ID
rs111298573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37677066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGGTGGCTCCTGACGCTAAGAG[C/G]TGAGAAGTGAAGTCGGGGTGGTGGG
Phenotype
MIM: 150570
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LGALS1 PubMed Links

Gene Details

Gene
LGALS1
Gene Name
galectin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002305.3 Intron NP_002296.1
Gene
NOL12
Gene Name
nucleolar protein 12
There are no transcripts associated with this gene.

View Full Product Details