Product Details

SNP ID
rs115499450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41694167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAAATCCAGGCATCTGGGGAGT[A/G]GTTCCCAGTAGACTTTGGAATGGAG
Phenotype
MIM: 608797 MIM: 601304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf46 PubMed Links

Gene Details

Gene
C22orf46
Gene Name
chromosome 22 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142964.1 1006 UTR 3 NP_001136436.1
XM_011530373.2 1006 UTR 3 XP_011528675.1
XM_017028939.1 1006 UTR 5 XP_016884428.1
Gene
MEI1
Gene Name
meiotic double-stranded break formation protein 1
There are no transcripts associated with this gene.

Gene
SNU13
Gene Name
SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5)
There are no transcripts associated with this gene.

View Full Product Details