Product Details
- SNP ID
-
rs115090509
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:46051840 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCGCGGTAGCTCATACCTGTAAT[G/T]CCAGCACATTGGGAGGCCGAGGCAG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00899
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59929908] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00899
- Gene Name
- long intergenic non-protein coding RNA 899
There are no transcripts associated with this gene.
- Gene
- PRR34
- Gene Name
- proline rich 34
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018280.2 |
455 |
Missense Mutation |
GAA,GCA |
E135A |
NP_060750.1 |
- Gene
- PRR34-AS1
- Gene Name
- PRR34 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details