Product Details

SNP ID
rs115112627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23148099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGATTCTGTGGCCAGGCTGGT[A/G]AGGAGGAGGTGACTCTAGACAGGTC
Phenotype
MIM: 605662 MIM: 605663
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB36 PubMed Links

Gene Details

Gene
RAB36
Gene Name
RAB36, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004914.3 Intron NP_004905.2
XM_005261859.4 Intron XP_005261916.1
XM_006724381.3 Intron XP_006724444.1
XM_006724382.3 Intron XP_006724445.1
XM_011530544.2 Intron XP_011528846.1
XM_011530545.2 Intron XP_011528847.1
XM_011530546.2 Intron XP_011528848.1
XM_011530547.2 Intron XP_011528849.1
XM_011530550.2 Intron XP_011528852.1
XM_011530552.2 Intron XP_011528854.1
XM_017029104.1 Intron XP_016884593.1
XM_017029105.1 Intron XP_016884594.1
XM_017029106.1 Intron XP_016884595.1
XM_017029107.1 Intron XP_016884596.1
Gene
RSPH14
Gene Name
radial spoke head 14 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014433.2 Intron NP_055248.1
XM_011530149.2 Intron XP_011528451.1
XM_011530150.1 Intron XP_011528452.1
XM_011530151.2 Intron XP_011528453.1
XM_011530152.2 Intron XP_011528454.1
XM_011530154.2 Intron XP_011528456.1
XM_011530155.2 Intron XP_011528457.1
XM_017028774.1 Intron XP_016884263.1

View Full Product Details