Product Details

SNP ID
rs104886492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53432410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTGCACATCCTTCTCAGAGGTC[G/A]CCTTCAAAGAGAGAAGTGGAGAAGG
Phenotype
MIM: 300256 MIM: 300040
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
HSD17B10 PubMed Links

Gene Details

Gene
HSD17B10
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037811.2 225 Missense Mutation GCG,GTG A65V NP_001032900.1
NM_004493.2 225 Missense Mutation GCG,GTG A65V NP_004484.1
Gene
RIBC1
Gene Name
RIB43A domain with coiled-coils 1
There are no transcripts associated with this gene.

Gene
SMC1A
Gene Name
structural maintenance of chromosomes 1A
There are no transcripts associated with this gene.

View Full Product Details