Product Details
- SNP ID
-
rs104894803
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:68829886 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAAGAACCTGGAGCCCGTATCCT[A/G]GAGCTCCCTCAACCCCAAGTGAGTA
- Phenotype
-
MIM: 300035
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFNB1
PubMed Links
Gene Details
- Gene
- EFNB1
- Gene Name
- ephrin B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004429.4 |
890 |
Nonsense Mutation |
TAG,TGG |
*37W |
NP_004420.1 |
View Full Product Details