Product Details

SNP ID
rs113706342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38796673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATTAAGCTATTAGATCCTTCA[T/G]GAAAGAGATTGTGATGGGCAAAGTA
Phenotype
MIM: 601194
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
TLR1 PubMed Links

Gene Details

Gene
TLR1
Gene Name
toll like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003263.3 2986 Missense Mutation CAT,CCT H720P NP_003254.2
XM_005262662.4 2986 Missense Mutation CAT,CCT H720P XP_005262719.1
XM_011513742.2 2986 Missense Mutation CAT,CCT H720P XP_011512044.1
XM_011513743.2 2986 Missense Mutation CAT,CCT H720P XP_011512045.1
XM_011513744.2 2986 Missense Mutation CAT,CCT H720P XP_011512046.1
XM_011513745.2 2986 Missense Mutation CAT,CCT H720P XP_011512047.1
XM_017008571.1 2986 Missense Mutation CAT,CCT H720P XP_016864060.1
XM_017008572.1 2986 Missense Mutation CAT,CCT H720P XP_016864061.1

View Full Product Details