Product Details

SNP ID
rs113069571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185019671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGATCCGCAGCCGCGTCCGCTC[G/T]CTGGTCCTCTCCAGCGCCACAGTGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HELT PubMed Links

Gene Details

Gene
HELT
Gene Name
helt bHLH transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300781.1 Intron NP_001287710.1
NM_001300782.1 Intron NP_001287711.1
XM_017008186.1 Intron XP_016863675.1

View Full Product Details