Product Details

SNP ID
rs116716724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:98878680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAGTTGGAAAATAACATATAGC[C/T]TAATTTTGTATTTAGAGAAGATGCT
Phenotype
MIM: 133440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF4E PubMed Links

Gene Details

Gene
EIF4E
Gene Name
eukaryotic translation initiation factor 4E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130678.1 4525 UTR 3 NP_001124150.1
NM_001130679.1 4525 UTR 3 NP_001124151.1
NM_001968.3 4525 UTR 3 NP_001959.1
XM_006714126.3 4525 Intron XP_006714189.1

View Full Product Details