Product Details

SNP ID
rs114395674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127794385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGTGTGCTACTATGTGACATCA[A/C]TTTTTCCCCAAATGGTCCCTCAGGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSPA4L PubMed Links

Gene Details

Gene
HSPA4L
Gene Name
heat shock protein family A (Hsp70) member 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317381.1 Intron NP_001304310.1
NM_001317382.1 Intron NP_001304311.1
NM_001317383.1 Intron NP_001304312.1
NM_014278.3 Intron NP_055093.2
XM_011531745.2 Intron XP_011530047.1

View Full Product Details