Product Details

SNP ID
rs112102623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154605069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCTTTTGAGTAAGTGCCACCT[A/T]AAACAAGTCGTAGATGTACATATCA
Phenotype
MIM: 134850
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FGG PubMed Links

Gene Details

Gene
FGG
Gene Name
fibrinogen gamma chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000509.5 Intron NP_000500.2
NM_021870.2 Intron NP_068656.2

View Full Product Details