Product Details

SNP ID
rs113731629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:170063078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGTCTGCTAAGACAAGAGGAGT[C/T]GGGAGTGATATGATTTGGAGTACTC
Phenotype
MIM: 611754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AADAT PubMed Links

Gene Details

Gene
AADAT
Gene Name
aminoadipate aminotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286682.1 Intron NP_001273611.1
NM_001286683.1 Intron NP_001273612.1
NM_016228.3 Intron NP_057312.1
NM_182662.1 Intron NP_872603.1
XM_006714231.2 Intron XP_006714294.1
XM_011532020.2 Intron XP_011530322.1

View Full Product Details